Order Entry
Czech Republic
ContactUsLinkComponent
(S)-1-[3-(Cyclopropyl-methyl-amino)-propyl]-1-(4-fluoro-phenyl)-1,3-dihydro-isobenzofuran-5-carbonitrile
(S)-1-[3-(Cyclopropyl-methyl-amino)-propyl]-1-(4-fluoro-phenyl)-1,3-dihydro-isobenzofuran-5-carbonitrile
Catalog # APOSPC408411-500MG
Supplier:  Apollo Scientific
CAS Number:  
(S)-1-[3-(Cyclopropyl-methyl-amino)-propyl]-1-(4-fluoro-phenyl)-1,3-dihydro-isobenzofuran-5-carbonitrile
Catalog # APOSPC408411-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :350.429 g/mol
Storage Temperature:Ambient
CAS Number:1421026-41-6
Formula:C₂₂H₂₃FN₂O
MDL Number:MFCD24369843