Order Entry
Czech Republic
ContactUsLinkComponent
3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ol
3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ol
Catalog # APOSPC408397-500MG
Supplier:  Apollo Scientific
CAS Number:  
3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ol
Catalog # APOSPC408397-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :280.674 g/mol
Storage Temperature:Ambient
CAS Number:1292498-24-8
Formula:C₁₁H₁₂ClF₃N₂O
MDL Number:MFCD13196948