Order Entry
Czech Republic
ContactUsLinkComponent
3-Piperidin-4-yl-2-trifluoromethyl-3H-imidazo[4,5-b]pyridine
3-Piperidin-4-yl-2-trifluoromethyl-3H-imidazo[4,5-b]pyridine
Catalog # APOSPC408396-500MG
Supplier:  Apollo Scientific
CAS Number:  
3-Piperidin-4-yl-2-trifluoromethyl-3H-imidazo[4,5-b]pyridine
Catalog # APOSPC408396-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :270.254 g/mol
Storage Temperature:Ambient
CAS Number:951544-48-2
Formula:C₁₂H₁₃F₃N₄
MDL Number:MFCD22631687