Order Entry
Czech Republic
ContactUsLinkComponent
(1R,4R)-N1-(4-Fluoro-benzyl)-cyclohexane-1,4-diamine hydrochloride
(1R,4R)-N1-(4-Fluoro-benzyl)-cyclohexane-1,4-diamine hydrochloride
Catalog # APOSPC408351-500MG
Supplier:  Apollo Scientific
CAS Number:  
(1R,4R)-N1-(4-Fluoro-benzyl)-cyclohexane-1,4-diamine hydrochloride
Catalog # APOSPC408351-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :258.763 g/mol
Storage Temperature:Ambient
CAS Number:1366386-71-1
Formula:C₁₃H₁₉FN₂·ClH
MDL Number:MFCD21098650