[1-(5-Fluoro-pyrimidin-2-yl)-piperidin-3-yl]-methanol
Catalog # APOSPC408312-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:211.236 g/mol
Storage Temperature:Ambient
CAS Number:1261232-82-9
Formula:C₁₀H₁₄FN₃O
MDL Number:MFCD18380078