3'-Chloro-5'-trifluoromethyl-3,4,5,6-tetrahydro-2H-[1,2']bipyridinyl-3-ylamine hydrochloride
Catalog # APOSPC408299-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:316.15 g/mol
Storage Temperature:Ambient
CAS Number:1185313-62-5
Formula:C₁₁H₁₃ClF₃N₃·ClH
MDL Number:MFCD09877740