2-Fluoro-5-iodo-4-methylpyridine 97%
Catalog # APOSPC408288-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:237.013 g/mol
Storage Temperature:Ambient
CAS Number:1184913-75-4
Formula:C₆H₅FIN
MDL Number:MFCD12911585