
2-Iodo-1,1,1,3,3,3-hexafluoropropane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4141-91-7
:
Formula:C₃HF₆I
MDL Number:MFCD00040984
:277.935 g/mol
Boiling Pt:58 °C
Density:2.11 g/cm³ (25 °C)
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together