Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,3,3-Trifluoro-1-phenylpropan-1-one
3,3,3-Trifluoro-1-phenylpropan-1-one 99%
3,3,3-Trifluoro-1-phenylpropan-1-one 99%
Irritant
3,3,3-Trifluoro-1-phenylpropan-1-one 99%
Catalog # APOSPC408142-5G
Supplier:
Apollo Scientific
CAS Number:
Expand image
Irritant
3,3,3-Trifluoro-1-phenylpropan-1-one 99%
Catalog # APOSPC408142-5G
Supplier:
Apollo Scientific
CAS Number:
About this item
Product Details
Product Details