(+/-)-3-(trans-2-(Trifluoromethyl)cyclopropyl)benzoic acid
Catalog # APOSPC405604-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:230.183 g/mol
Storage Temperature:Ambient
CAS Number:1404192-15-9
Formula:C₁₁H₉F₃O₂
MDL Number:MFCD22683236