Order Entry
Czech Republic
ContactUsLinkComponent
1-(Trifluoromethyl)-2-thia-6-azaspiro[3.3]heptane hemioxalate
1-(Trifluoromethyl)-2-thia-6-azaspiro[3.3]heptane hemioxalate
Catalog # APOSPC405521-250MG
Supplier:  Apollo Scientific
CAS Number:  
1-(Trifluoromethyl)-2-thia-6-azaspiro[3.3]heptane hemioxalate
Catalog # APOSPC405521-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:1980054-87-2
MDL Number:MFCD28009691