Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
1-(Trifluoromethyl)-2-oxa-6-azaspiro[3.3]heptane hemioxalate
Catalog # APOSPC405520-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(Trifluoromethyl)-2-oxa-6-azaspiro[3.3]heptane hemioxalate
Catalog # APOSPC405520-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :GHS07
Storage Temperature:Freezer
CAS Number:1454909-15-9
MDL Number:MFCD28009690