Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Fluorocyclobutanamine hydrochloride
3-Fluorocyclobutanamine hydrochloride
3-Fluorocyclobutanamine hydrochloride
3-Fluorocyclobutanamine hydrochloride
APOSPC405503-250MG
:
Apollo Scientific
:
3-Fluorocyclobutanamine hydrochloride
APOSPC405503-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
125.572 g/mol
:
GHS07
:
Ambient
:
1284245-36-8
:
C₄H₈FN·ClH
:
MFCD21642364