Order Entry
Czech Republic
ContactUsLinkComponent
5,7-Difluoro-4-hydroxy-2-methylquinoline
5,7-Difluoro-4-hydroxy-2-methylquinoline
Catalog # APOSPC404587-1G
Supplier:  Apollo Scientific
CAS Number:  
5,7-Difluoro-4-hydroxy-2-methylquinoline
Catalog # APOSPC404587-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.165 g/mol
Storage Temperature:Ambient
CAS Number:288151-40-6
Formula:C₁₀H₇F₂NO
MDL Number:MFCD11501289