4-Bromo-2-methyl-6-trifluoromethoxyquinoline
Catalog # APOSPC404586-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:306.079 g/mol
Storage Temperature:Ambient
CAS Number:1189107-42-3
Formula:C₁₁H₇BrF₃NO
MDL Number:MFCD12675022