4-Bromo-2-methyl-8-trifluoromethylquinoline
Catalog # APOSPC404573-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:290.079 g/mol
Storage Temperature:Ambient
CAS Number:1070879-58-1
Formula:C₁₁H₇BrF₃N
MDL Number:MFCD11505142