4-Bromo-8-fluoro-2-methylquinoline
Catalog # APOSPC404572-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:240.072 g/mol
Storage Temperature:Ambient
CAS Number:1070879-49-0
Formula:C₁₀H₇BrFN
MDL Number:MFCD11505134