4-Bromo-6-trifluoromethylquinoline
Catalog # APOSPC404569-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:276.053 g/mol
Storage Temperature:Ambient
CAS Number:1070879-32-1
Formula:C₁₀H₅BrF₃N
MDL Number:MFCD11505113