2-Chloro-6-fluoroquinoline-3-carbonitrile
Catalog # APOSPC404557-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:206.604 g/mol
Storage Temperature:Ambient
CAS Number:948291-71-2
Formula:C₁₀H₄ClFN₂
MDL Number:MFCD09787672