2,8-Bis(trifluoromethyl)-2'-(2-pyridyl)-4-quinolineacetonitrile
Catalog # APOSPC404550-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:381.275 g/mol
Storage Temperature:Ambient
CAS Number:83012-12-8
Formula:C₁₈H₉F₆N₃
MDL Number:MFCD08437551