6-(2,8-Bis-trifluoromethyl-quinolin-4-ylmethyl)-pyridin-3-ol
Catalog # APOSPC404549-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:372.265 g/mol
Storage Temperature:Ambient
CAS Number:955315-33-0
Formula:C₁₇H₁₀F₆N₂O
MDL Number:MFCD05863553