1-(4-(Trifluoromethoxy)phenyl)cyclopropanecarbonitrile
Catalog # APOSPC404540-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:227.183 g/mol
Storage Temperature:Ambient
CAS Number:173859-39-7
Formula:C₁₁H₈F₃NO
MDL Number:MFCD11036656