1-a-Amino(2-hydroxyphenyl)methyl-2-naphthol trifluoroacetate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:916825-05-3
:
Formula:C₁₇H₁₅NO₂·CF₃CO₂H
MDL Number:MFCD09881437
:379.33 g/mol
Storage Temperature:Ambient
Frequently Bought Together