Order Entry
Czech Republic
ContactUsLinkComponent
1-(4-Fluorophenyl)-1H-pyrazole-4-carboxylic acid 95%
1-(4-Fluorophenyl)-1H-pyrazole-4-carboxylic acid 95%
Catalog # APOSPC403055-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Fluorophenyl)-1H-pyrazole-4-carboxylic acid 95%
Catalog # APOSPC403055-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :206.173 g/mol
Storage Temperature:Ambient
CAS Number:138907-81-0
Formula:C₁₀H₇FN₂O₂
MDL Number:MFCD07186399