Order Entry
Czech Republic
ContactUsLinkComponent
1-(2-Fluorophenyl)-1H-pyrazole-4-carboxylic acid 95+%
1-(2-Fluorophenyl)-1H-pyrazole-4-carboxylic acid 95+%
Catalog # APOSPC403053-250MG
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Fluorophenyl)-1H-pyrazole-4-carboxylic acid 95+%
Catalog # APOSPC403053-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :206.173 g/mol
Storage Temperature:Ambient
CAS Number:1134310-63-6
Formula:C₁₀H₇FN₂O₂
MDL Number:MFCD09950609