2-[3-(Trifluoromethyl)phenyl]propan-2-amine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:153390-61-5
Formula:C₁₀H₁₂F₃N·ClH
MDL Number:MFCD14708148
:239.665 g/mol
Storage Temperature:Ambient
Frequently Bought Together