1-(4-Fluorophenyl)piperidine-2-carboxylic acid
Catalog # APOSPC402138-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:223.243 g/mol
Storage Temperature:Refrigerator
CAS Number:1504689-74-0
Formula:C₁₂H₁₄FNO₂
MDL Number:MFCD22407916