3-(4-(Trifluoromethyl)phenyl)piperidine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1106940-96-8
:
Formula:C₁₂H₁₄F₃N·ClH
MDL Number:MFCD02178940
:265.702 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together