4-(2-(Trifluoromethyl)phenyl)-1,2,3,6-tetrahydropyridine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:208989-34-8
:
Formula:C₁₂H₁₂F₃N·ClH
MDL Number:MFCD22378606
:263.687 g/mol
Storage Temperature:Ambient
Frequently Bought Together