0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
(8-Fluoroquinolin-3-yl)boronic acid
(8-Fluoroquinolin-3-yl)boronic acid 95%
(8-Fluoroquinolin-3-yl)boronic acid 95%
APOSPC402116-1G
:
Apollo Scientific
:
(8-Fluoroquinolin-3-yl)boronic acid 95%
APOSPC402116-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
190.967 g/mol
:
GHS07
:
Refrigerator
:
1207750-07-9
:
C₉H₇BFNO₂
:
MFCD18382879