Order Entry
Czech Republic
ContactUsLinkComponent
3-Bromo-2-(chloromethyl)-6-fluoropyridine 97%
Irritant
3-Bromo-2-(chloromethyl)-6-fluoropyridine 97%
Catalog # APOSPC402113-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Bromo-2-(chloromethyl)-6-fluoropyridine 97%
Catalog # APOSPC402113-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :224.458 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1227496-41-4
Formula:C₆H₄BrClFN
MDL Number:MFCD16608188