
3,5-Difluoro-3'-(trifluoromethoxy)-[1,1'-biphenyl]-4-amine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:867288-01-5
:
Formula:C₁₃H₈F₅NO
MDL Number:MFCD18400452
:289.201 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together