Order Entry
Czech Republic
ContactUsLinkComponent
8-(Trifluoromethyl)-1,4-dioxaspiro[4.5]decan-8-ol
Irritant
8-(Trifluoromethyl)-1,4-dioxaspiro[4.5]decan-8-ol
Katalogové číslo APOSPC402041-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
8-(Trifluoromethyl)-1,4-dioxaspiro[4.5]decan-8-ol
Katalogové číslo APOSPC402041-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :226.193 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1248081-37-9
Formula:C₉H₁₃F₃O₃
MDL Number:MFCD16067897