[5-(Trifluoromethyl)pyridin-2-yl]acetonitrile
Catalog # APOSPC402036-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:186.134 g/mol
Storage Temperature:Ambient
CAS Number:939793-18-7
Formula:C₈H₅F₃N₂
MDL Number:MFCD08690162