
4-(Dimethylamino)-3-(trifluoroacetyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:230305-72-3
:
Formula:C₉H₉F₃N₂O
MDL Number:MFCD09904820
Boiling Pt:70 °C (4 mm Hg)
:218.176 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together