2-Chloro-1-(2,6-dichloro-3-fluorophenyl)ethanone
Catalog # APOSPC401501-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:241.474 g/mol
Storage Temperature:Ambient
CAS Number:1249849-11-3
Formula:C₈H₄Cl₃FO
MDL Number:MFCD16093805