Ethyl 3-amino-6-(trifluoromethyl)thieno[3,2-b]pyridine-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1357943-66-8
:
Formula:C₁₁H₉F₃N₂O₂S
MDL Number:MFCD21364902
:290.262 g/mol
Storage Temperature:Ambient
Frequently Bought Together