Order Entry
Czech Republic
ContactUsLinkComponent
(2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(dimethylamino)prop-2-enal
(2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(dimethylamino)prop-2-enal
Catalog # APOSPC401049-1G
Supplier:  Apollo Scientific
CAS Number:  
(2-[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(dimethylamino)prop-2-enal
Catalog # APOSPC401049-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :278.658 g/mol
Storage Temperature:Ambient
CAS Number:339010-44-5
Formula:C₁₁H₁₀ClF₃N₂O
MDL Number:MFCD00231994