3-[Bis(2-hydroxyethyl)amino]-2-cyano-5-(trifluoromethyl)pyridine
Catalog # APOSPC401038-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:275.227 g/mol
Storage Temperature:Ambient
CAS Number:1672655-86-5
Formula:C₁₁H₁₂F₃N₃O₂
MDL Number:MFCD28122855