
6,8-Dihydro-2-[4-(trifluoromethyl)phenyl]-5H-thiopyrano[3,4-d]pyrimidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1923237-05-1
:
Formula:C₁₄H₁₁F₃N₂S
MDL Number:MFCD24369740
:296.311 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together