5-Fluoro-2-Methyl-3-nitrobenzoic acid
Catalog # APOSPC400720-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:199.136 g/mol
Storage Temperature:Ambient
CAS Number:850462-64-5
Formula:C₈H₆FNO₄
MDL Number:MFCD12913348