2,2,2-Trifluoroacetic acid compound with 3-(2-(methylamino)ethyl)benzoic acid
Catalog # APOSPC400598-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:293.239 g/mol
Storage Temperature:Ambient
CAS Number:1624261-41-1
Formula:C₁₀H₁₃NO₂·CF₃CO₂H
MDL Number:MFCD27978439