2-Fluoro-2-phenylcyclopropanecarboxylic acid
Catalog # APOSPC400596-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:180.176 g/mol
Storage Temperature:Ambient
CAS Number:914221-42-4
Formula:C₁₀H₉FO₂
MDL Number:MFCD09030645