(2,2-Dichloro-1-fluorocyclopropyl)benzene
Catalog # APOSPC400566-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:205.056 g/mol
Storage Temperature:Ambient
CAS Number:502763-69-1
Formula:C₉H₇Cl₂F
MDL Number:MFCD27501036