
2-(2-Aminoethyl)-3-chloro-5-(trifluoromethyl)pyridine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:658066-44-5
:
Formula:C₈H₈ClF₃N₂
MDL Number:MFCD11848345
:224.611 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together