
Methyl 5,5-difluoropiperidine-3-carboxylate, N-BOC protected
Katalogové číslo APOSPC400520-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:279.28 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1255667-06-1
Formula:C₁₂H₁₉F₂NO₄
MDL Number:MFCD18433629