N'-[4-(Trifluoromethoxy)benzyl]ethane-1,2-diamine, N-BOC protected
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:934757-43-4
:
Formula:C₁₅H₂₁F₃N₂O₃
MDL Number:MFCD22123822
:334.334 g/mol
Storage Temperature:Ambient
Frequently Bought Together