
(4-(Difluoromethoxy)pyridin-2-yl)methanol
Catalog # APOSPC400115-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:175.133 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1315361-55-7
Formula:C₇H₇F₂NO₂
MDL Number:MFCD22552054