

1-(2,2-Dibromovinyl)-4-chloro-2-fluorobenzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1862532-60-2
:
Formula:C₈H₄Br₂ClF
MDL Number:MFCD31561227
:314.377 g/mol
UN:2811
ADR:6.1,III
:GHS05
:GHS06
Storage Temperature:Ambient
Frequently Bought Together