0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
2-(2-Bromoethynyl)-1-chloro-3-fluorobenzene
2-(2-Bromoethynyl)-1-chloro-3-fluorobenzene
2-(2-Bromoethynyl)-1-chloro-3-fluorobenzene
APOSPC400028-250MG
:
Apollo Scientific
:
2-(2-Bromoethynyl)-1-chloro-3-fluorobenzene
APOSPC400028-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
233.465 g/mol
:
GHS07
:
Freezer
:
2227272-53-7
:
C₈H₃BrClF
:
MFCD31536746